Assay Details
| Gene Symbol: Gfy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066420.2 | XP_012921874.1 | 3 - 4 | 2325 | 98 | |
| XM_021246052.1 | XP_021101711.1 | 4 - 5 | 2445 | 98 | ||
More Information
Set Membership: |
|

Gfy
Genomic Map