Assay Details
| Gene Symbol: Osgin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841965.3 | XP_004842022.2 | 2 - 3 | 420 | 79 | |
| XM_013071195.2 | XP_012926649.1 | 2 - 3 | 555 | 79 | ||
| XM_021255965.1 | XP_021111624.1 | 2 - 3 | 178 | 79 | ||
More Information
Set Membership: |
|

Osgin2
Genomic Map