Assay Details
| Gene Symbol: Rgs8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862826.2 | XP_004862883.1 | 6 - 7 | 614 | 76 | |
| XM_004862827.3 | XP_004862884.1 | 5 - 6 | 621 | 76 | ||
| XM_004862828.2 | XP_004862885.1 | 5 - 6 | 469 | 76 | ||
| XM_013078938.2 | XP_012934392.1 | 5 - 6 | 640 | 76 | ||
| XM_021242901.1 | XP_021098560.1 | 6 - 7 | 750 | 76 | ||
| XM_021242902.1 | XP_021098561.1 | 6 - 7 | 558 | 76 | ||
| XM_021242903.1 | XP_021098562.1 | 5 - 6 | 610 | 76 | ||
| XM_021242904.1 | XP_021098563.1 | 5 - 6 | 459 | 76 | ||
| XM_021242905.1 | XP_021098564.1 | 5 - 6 | 618 | 76 | ||
More Information
Set Membership: |
|

Rgs8
Genomic Map