Assay Details
| Gene Symbol: Ccdc120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871982.3 | XP_004872039.1 | 6 - 7 | 2220 | 55 | |
| XM_013068227.2 | XP_012923681.1 | 6 - 7 | 2226 | 55 | ||
| XM_013068228.2 | XP_012923682.1 | 6 - 7 | 2240 | 55 | ||
| XM_013068229.2 | XP_012923683.1 | 5 - 6 | 874 | 55 | ||
| XM_021249973.1 | XP_021105632.1 | 6 - 7 | 1015 | 55 | ||
| XM_021249974.1 | XP_021105633.1 | 5 - 6 | 2085 | 55 | ||
More Information
Set Membership: |
|

Ccdc120
Genomic Map