Assay Details
| Gene Symbol: Ttc23l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261514.1 | XP_021117173.1 | 7 - 8 | 923 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc23l| Gene Symbol: Ttc23l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021261514.1 | XP_021117173.1 | 7 - 8 | 923 | 72 | |
Set Membership: |
|