Assay Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843949.3 | XP_004844006.1 | 2 - 3 | 599 | 58 | |
| XM_021257612.1 | XP_021113271.1 | 2 - 3 | 607 | 58 | ||
| XM_021257613.1 | XP_021113272.1 | 2 - 3 | 500 | 58 | ||
More Information
Set Membership: |
|

Rilpl2
Genomic Map