Assay Details
| Gene Symbol: Epyc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844981.2 | XP_004845038.1 | 6 - 7 | 881 | 70 | |
| XM_021258487.1 | XP_021114146.1 | 6 - 7 | 915 | 70 | ||
More Information
Set Membership: |
|

Epyc
Genomic Map