Assay Details
| Gene Symbol: Mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873224.3 | XP_004873281.1 | 5 - 6 | 618 | 74 | |
| XM_021251098.1 | XP_021106757.1 | 5 - 6 | 618 | 74 | ||
| XM_021251099.1 | XP_021106758.1 | 4 - 5 | 604 | 74 | ||
| XM_021251100.1 | XP_021106759.1 | 7 - 8 | 1278 | 74 | ||
| XM_021251102.1 | XP_021106761.1 | 8 - 9 | 1208 | 74 | ||
| XM_021251103.1 | XP_021106762.1 | 6 - 7 | 1023 | 74 | ||
More Information
Set Membership: |
|

Mvb12a
Genomic Map