Assay Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847032.3 | XP_004847089.1 | 2 - 3 | 683 | 94 | |
| XM_004847034.3 | XP_004847091.1 | 2 - 3 | 685 | 94 | ||
| XM_013072779.2 | XP_012928233.1 | 2 - 3 | 684 | 94 | ||
More Information
Set Membership: |
|

Tcf19
Genomic Map