Assay Details
| Gene Symbol: Tcf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847032.3 | XP_004847089.1 | 3 - 4 | 1228 | 95 | |
| XM_013072779.2 | XP_012928233.1 | 3 - 4 | 1229 | 95 | ||
More Information
Set Membership: |
|

Tcf19
Genomic Map