Assay Details
| Gene Symbol: Snx20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847942.2 | XP_004847999.1 | 3 - 4 | 412 | 64 | |
| XM_004847943.2 | XP_004848000.1 | 3 - 4 | 415 | 64 | ||
| XM_004847945.2 | XP_004848002.1 | 2 - 3 | 273 | 64 | ||
| XM_021260929.1 | XP_021116588.1 | 4 - 5 | 523 | 64 | ||
More Information
Set Membership: |
|

Snx20
Genomic Map