Assay Details
| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849778.2 | XP_004849835.1 | 3 - 4 | 453 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrg1| Gene Symbol: Gabrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849778.2 | XP_004849835.1 | 3 - 4 | 453 | 77 | |
Set Membership: |
|