Assay Details
| Gene Symbol: Znf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068584.2 | XP_012924038.1 | 3 - 4 | 187 | 63 | |
| XM_013068585.2 | XP_012924039.1 | 4 - 5 | 325 | 63 | ||
| XM_013068586.2 | XP_012924040.1 | 2 - 3 | 142 | 63 | ||
| XM_013068587.2 | XP_012924041.1 | 2 - 3 | 332 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013068584.2, XM_013068585.2, XM_013068586.2, XM_013068587.2 transcripts.
Set Membership: |
|

Znf45
Genomic Map