Assay Details
| Gene Symbol: Znf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068584.2 | XP_012924038.1 | 4 - 5 | 309 | 103 | |
| XM_013068585.2 | XP_012924039.1 | 5 - 6 | 447 | 103 | ||
| XM_013068586.2 | XP_012924040.1 | 3 - 4 | 264 | 103 | ||
| XM_013068587.2 | XP_012924041.1 | 3 - 4 | 454 | 103 | ||
More Information
Set Membership: |
|

Znf45
Genomic Map