Assay Details
| Gene Symbol: Tex35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075341.2 | XP_012930795.1 | 6 - 7 | 505 | 116 | |
| XM_013075342.2 | XP_012930796.1 | 6 - 7 | 505 | 116 | ||
| XM_013075343.2 | XP_012930797.1 | 6 - 7 | 505 | 116 | ||
| XM_013075344.2 | XP_012930798.1 | 6 - 7 | 505 | 116 | ||
| XM_021265368.1 | XP_021121027.1 | 6 - 7 | 505 | 116 | ||
| XM_021265369.1 | XP_021121028.1 | 6 - 7 | 505 | 116 | ||
| XM_021265370.1 | XP_021121029.1 | 6 - 7 | 505 | 116 | ||
| XM_021265371.1 | XP_021121030.1 | 6 - 7 | 505 | 116 | ||
| XM_021265372.1 | XP_021121031.1 | 5 - 6 | 445 | 116 | ||
| XM_021265373.1 | XP_021121032.1 | 5 - 6 | 436 | 116 | ||
| XM_021265374.1 | XP_021121033.1 | 5 - 6 | 445 | 116 | ||
| XM_021265375.1 | XP_021121034.1 | 4 - 5 | 376 | 116 | ||
More Information
Set Membership: |
|

Tex35
Genomic Map