Assay Details
| Gene Symbol: Spata19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862659.3 | XP_004862716.1 | 5 - 6 | 499 | 68 | |
| XM_021242785.1 | XP_021098444.1 | 4 - 5 | 410 | 68 | ||
More Information
Set Membership: |
|

Spata19
Genomic Map