Assay Details
| Gene Symbol: Hps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834421.3 | XP_004834478.1 | 9 - 10 | 1788 | 54 | |
| XM_004834422.3 | XP_004834479.1 | 9 - 10 | 1788 | 54 | ||
| XM_004834424.2 | XP_004834481.1 | 10 - 11 | 1673 | 54 | ||
More Information
Set Membership: |
|

Hps3
Genomic Map