Assay Details
| Gene Symbol: Pcgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866518.3 | XP_004866575.1 | 5 - 6 | 972 | 115 | |
| XM_021245568.1 | XP_021101227.1 | 5 - 6 | 980 | 115 | ||
| XM_021245569.1 | XP_021101228.1 | 5 - 6 | 984 | 115 | ||
| XM_021245570.1 | XP_021101229.1 | 5 - 6 | 981 | 115 | ||
| XM_021245572.1 | XP_021101231.1 | 5 - 6 | 984 | 115 | ||
| XM_021245573.1 | XP_021101232.1 | 5 - 6 | 981 | 115 | ||
| XM_021245574.1 | XP_021101233.1 | 5 - 6 | 983 | 115 | ||
| XM_021245575.1 | XP_021101234.1 | 5 - 6 | 978 | 115 | ||
More Information
Set Membership: |
|

Pcgf6
Genomic Map