Assay Details
| Gene Symbol: Tmc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860810.3 | XP_004860867.1 | 7 - 8 | 2984 | 58 | |
| XM_021241165.1 | XP_021096824.1 | 7 - 8 | 2984 | 58 | ||
| XM_021241166.1 | XP_021096825.1 | 7 - 8 | 2984 | 58 | ||
| XM_021241167.1 | XP_021096826.1 | 8 - 9 | 2708 | 58 | ||
| XM_021241168.1 | XP_021096827.1 | 7 - 8 | 1004 | 58 | ||
More Information
Set Membership: |
|

Tmc8
Genomic Map