Assay Details
| Gene Symbol: Gmfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835510.2 | XP_004835567.1 | 3 - 4 | 271 | 74 | |
| XM_021262093.1 | XP_021117752.1 | 3 - 4 | 271 | 74 | ||
More Information
Set Membership: |
|

Gmfb
Genomic Map