Assay Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849197.3 | XP_004849254.1 | 4 - 5 | 711 | 60 | |
| XM_021261922.1 | XP_021117581.1 | 4 - 5 | 708 | 60 | ||
| XM_021261923.1 | XP_021117582.1 | 4 - 5 | 711 | 60 | ||
More Information
Set Membership: |
|

Wdr34
Genomic Map