Assay Details
| Gene Symbol: Lrrc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841306.3 | XP_004841363.1 | 3 - 4 | 316 | 124 | |
| XM_013070896.2 | XP_012926350.1 | 4 - 5 | 359 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004841306.3, XM_013070896.2 transcripts.
Set Membership: |
|

Lrrc8b
Genomic Map