Assay Details
| Gene Symbol: Lrrc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841306.3 | XP_004841363.1 | 4 - 5 | 2483 | 62 | |
| XM_013070896.2 | XP_012926350.1 | 5 - 6 | 2526 | 62 | ||
More Information
Set Membership: |
|

Lrrc8b
Genomic Map