Assay Details
| Gene Symbol: Uhrf1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846733.3 | XP_004846790.1 | 6 - 7 | 872 | 61 | |
| XM_021259964.1 | XP_021115623.1 | 6 - 7 | 872 | 61 | ||
| XM_021259965.1 | XP_021115624.1 | 6 - 7 | 807 | 61 | ||
| XM_021259967.1 | XP_021115626.1 | 6 - 7 | 649 | 61 | ||
| XM_021259968.1 | XP_021115627.1 | 6 - 7 | 874 | 61 | ||
More Information
Set Membership: |
|

Uhrf1bp1
Genomic Map