Assay Details
| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262117.1 | XP_021117776.1 | 2 - 3 | 1723 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb26| Gene Symbol: Zbtb26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262117.1 | XP_021117776.1 | 2 - 3 | 1723 | 77 | |
Set Membership: |
|