Assay Details
| Gene Symbol: Has1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866752.3 | XP_004866809.1 | 3 - 4 | 1022 | 60 | |
| XM_021246265.1 | XP_021101924.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246266.1 | XP_021101925.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246267.1 | XP_021101926.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246268.1 | XP_021101927.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246269.1 | XP_021101928.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246270.1 | XP_021101929.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246271.1 | XP_021101930.1 | 2 - 3 | 1483 | 60 | ||
| XM_021246272.1 | XP_021101931.1 | 3 - 4 | 935 | 60 | ||
More Information
Set Membership: |
|

Has1
Genomic Map