Assay Details
| Gene Symbol: Opn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867722.2 | XP_004867779.1 | 1 - 2 | 1290 | 55 | |
| XM_021246766.1 | XP_021102425.1 | 1 - 2 | 1290 | 55 | ||
| XM_021246767.1 | XP_021102426.1 | 1 - 2 | 1290 | 55 | ||
More Information
Set Membership: |
|

Opn4
Genomic Map