Assay Details
| Gene Symbol: Mpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013067643.2 | XP_012923097.1 | 4 - 5 | 1022 | 51 | |
| XM_021248715.1 | XP_021104374.1 | 3 - 4 | 1132 | 51 | ||
More Information
Set Membership: |
|

Mpo
Genomic Map