Assay Details
| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845068.3 | XP_004845125.1 | 4 - 5 | 551 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nuak1| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845068.3 | XP_004845125.1 | 4 - 5 | 551 | 66 | |
Set Membership: |
|