Assay Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851207.2 | XP_004851264.1 | 7 - 8 | 972 | 64 | |
| XM_021263624.1 | XP_021119283.1 | 7 - 8 | 974 | 64 | ||
| XM_021263625.1 | XP_021119284.1 | 7 - 8 | 986 | 64 | ||
| XM_021263626.1 | XP_021119285.1 | 6 - 7 | 914 | 64 | ||
More Information
Set Membership: |
|

Cdk15
Genomic Map