Assay Details
| Gene Symbol: Trpc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861420.2 | XP_004861477.1 | 10 - 11 | 2556 | 68 | |
| XM_004861421.2 | XP_004861478.1 | 10 - 11 | 2559 | 68 | ||
| XM_021241600.1 | XP_021097259.1 | 10 - 11 | 2542 | 68 | ||
More Information
Set Membership: |
|

Trpc5
Genomic Map