Assay Details
| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863817.3 | XP_004863874.2 | 3 - 4 | 369 | 58 | |
| XM_013065182.2 | XP_012920636.1 | 3 - 4 | 1161 | 58 | ||
| XM_013065183.2 | XP_012920637.1 | 3 - 4 | 1115 | 58 | ||
| XM_021243541.1 | XP_021099200.1 | 4 - 5 | 502 | 58 | ||
| XM_021243542.1 | XP_021099201.1 | 4 - 5 | 456 | 58 | ||
More Information
Set Membership: |
|

Gabrd
Genomic Map