Assay Details
| Gene Symbol: Chst9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013075285.2 | XP_012930739.1 | 4 - 5 | 439 | 88 | |
| XM_021265127.1 | XP_021120786.1 | 5 - 6 | 478 | 88 | ||
More Information
Set Membership: |
|

Chst9
Genomic Map