Assay Details
| Gene Symbol: Tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835543.3 | XP_004835600.1 | 4 - 5 | 525 | 70 | |
| XM_004835544.3 | XP_004835601.1 | 4 - 5 | 546 | 70 | ||
| XM_004835545.3 | XP_004835602.1 | 4 - 5 | 437 | 70 | ||
More Information
Set Membership: |
|

Tmx1
Genomic Map