Assay Details
| Gene Symbol: Padi6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850391.2 | XP_004850448.1 | 15 - 16 | 1897 | 61 | |
| XM_021262910.1 | XP_021118569.1 | 15 - 16 | 1912 | 61 | ||
| XM_021262911.1 | XP_021118570.1 | 15 - 16 | 1816 | 61 | ||
More Information
Set Membership: |
|

Padi6
Genomic Map