Assay Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856732.3 | XP_004856789.2 | 7 - 8 | 1215 | 63 | |
| XM_004856733.3 | XP_004856790.1 | 7 - 8 | 1206 | 63 | ||
| XM_004856734.3 | XP_004856791.1 | 6 - 7 | 1102 | 63 | ||
| XM_004856735.3 | XP_004856792.1 | 5 - 6 | 1044 | 63 | ||
| XM_004856736.3 | XP_004856793.1 | 4 - 5 | 672 | 63 | ||
| XM_021237908.1 | XP_021093567.1 | 6 - 7 | 1148 | 63 | ||
| XM_021237909.1 | XP_021093568.1 | 5 - 6 | 826 | 63 | ||
More Information
Set Membership: |
|

Bcl9l
Genomic Map