Assay Details
| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870157.3 | XP_004870214.3 | 3 - 4 | 395 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sit1| Gene Symbol: Sit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870157.3 | XP_004870214.3 | 3 - 4 | 395 | 57 | |
Set Membership: |
|