Assay Details
| Gene Symbol: Opa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867145.3 | XP_004867202.1 | 2 | 420 | 93 | |
| XM_021245817.1 | XP_021101476.1 | 3 | 949 | 93 | ||
More Information
Set Membership: |
|

Opa3
Genomic Map