Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000499.4 | NP_000490.1 | 1 - 2 | 168 | 91 | |
| GenBank mRNA | AK223113.1 | - | 1 - 2 | 86 | 91 | |
| AK313880.1 | - | 1 - 2 | 86 | 91 | ||
| K03191.1 | - | 1 - 2 | 58 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001647.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313880.1,K03191.1,NM_000499.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
porphyrin-containing compound metabolic process
aging
cell proliferation
amine metabolic process
response to wounding
response to virus
response to nematode
response to herbicide
ethylene metabolic process
embryo development ending in birth or egg hatching
coumarin metabolic process
flavonoid metabolic process
response to iron(III) ion
insecticide metabolic process
drug metabolic process
dibenzo-p-dioxin catabolic process
epoxygenase P450 pathway
response to food
response to lipopolysaccharide
response to vitamin A
response to immobilization stress
vitamin D metabolic process
response to drug
9-cis-retinoic acid biosynthetic process
camera-type eye development
response to antibiotic
response to arsenic-containing substance
digestive tract development
hydrogen peroxide biosynthetic process
response to hyperoxia
oxidation-reduction process
maternal process involved in parturition
hepatocyte differentiation
demethylation
cellular response to organic cyclic compound
omega-hydroxylase P450 pathway
positive regulation of G1/S transition of mitotic cell cycle
monooxygenase activity
iron ion binding
protein binding
steroid hydroxylase activity
oxidoreductase activity
oxidoreductase activity, acting on diphenols and related substances as donors
flavonoid 3'-monooxygenase activity
oxygen binding
enzyme binding
heme binding
demethylase activity
aromatase activity
vitamin D 24-hydroxylase activity
response to hypoxia
porphyrin-containing compound metabolic process
aging
cell proliferation
amine metabolic process
response to wounding
response to virus
response to nematode
response to herbicide
ethylene metabolic process
embryo development ending in birth or egg hatching
coumarin metabolic process
flavonoid metabolic process
response to iron(III) ion
insecticide metabolic process
drug metabolic process
dibenzo-p-dioxin catabolic process
epoxygenase P450 pathway
response to food
response to lipopolysaccharide
response to vitamin A
response to immobilization stress
vitamin D metabolic process
response to drug
9-cis-retinoic acid biosynthetic process
camera-type eye development
response to antibiotic
response to arsenic-containing substance
digestive tract development
hydrogen peroxide biosynthetic process
response to hyperoxia
oxidation-reduction process
maternal process involved in parturition
hepatocyte differentiation
demethylation
cellular response to organic cyclic compound
omega-hydroxylase P450 pathway
positive regulation of G1/S transition of mitotic cell cycle
monooxygenase activity
iron ion binding
protein binding
steroid hydroxylase activity
oxidoreductase activity
oxidoreductase activity, acting on diphenols and related substances as donors
flavonoid 3'-monooxygenase activity
oxygen binding
enzyme binding
heme binding
demethylase activity
aromatase activity
vitamin D 24-hydroxylase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000379727.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395048.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000395049.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000562201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000564596.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000566503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000567032.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000569630.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000612821.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | ENST00000617691.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010042.hg.1 | NM_000499.4 | Refseq |
Citations (1)
Altered gene expression patterns in MCF-7 cells induced by the urban dust particulate complex mixture standard reference material 1649a.
- Authors
- Mahadevan B, Keshava C, Musafia-Jeknic T, Pecaj A, Weston A, Baird WM
- Journal
- Cancer Res - Page No 1251 (Tue Feb 15 00:00:00 UTC 2005)
- Volume (Year)
- 65 (2005)
- Product Usage
- -

CYP1A1
Genomic Map