| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017608.hg.1 |
ENST00000395743.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017608.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017608.hg.1 |
TC04001292.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017609.hg.1 |
ENST00000395743.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017609.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017609.hg.1 |
TC04001292.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017607.hg.1 |
ENST00000395743.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017607.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017607.hg.1 |
TC04001292.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400011027.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
ENST00000512743.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174959.hg.1 |
TC0400011027.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174960.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174960.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174960.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174960.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174960.hg.1 |
TC0400011027.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174962.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174962.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174962.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174962.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174962.hg.1 |
TC0400011027.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400174961.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174961.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174961.hg.1 |
ENST00000395743.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400174961.hg.1 |
NM_001729.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400174961.hg.1 |
TC0400011027.hg.1 |
Clariom D TC |