Assay Details
| Gene Symbol: XPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000380.3 | NP_000371.1 | 1 - 2 | 288 | 91 | |
| NR_027302.1 | 1 - 2 | 288 | 91 | |||
| XM_006717278.1 | XP_006717341.1 | 1 - 2 | 282 | 91 | ||
| GenBank mRNA | AL531629.3 | - | 1 - 2 | 191 | 91 | |
| BC014965.1 | - | 1 - 2 | 236 | 4878739 | 91 | |
| BT019518.1 | - | 1 - 2 | 171 | 91 | ||
| CB155090.1 | - | 1 - 2 | 288 | 91 | ||
| D14533.1 | - | 1 - 2 | 197 | 91 | ||
| KC899693.1 | - | 1 - 2 | 171 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001397.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleotide-excision repair, DNA damage recognition
nucleotide-excision repair, DNA duplex unwinding
DNA repair
transcription-coupled nucleotide-excision repair
base-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
response to oxidative stress
intrinsic apoptotic signaling pathway in response to DNA damage
response to toxic substance
response to auditory stimulus
nucleotide-excision repair, DNA incision
multicellular organism growth
global genome nucleotide-excision repair
UV-damage excision repair
nucleotide-excision repair involved in interstrand cross-link repair
damaged DNA binding
protein binding
protein domain specific binding
protein homodimerization activity
metal ion binding
nucleotide-excision repair, DNA damage recognition
nucleotide-excision repair, DNA duplex unwinding
DNA repair
transcription-coupled nucleotide-excision repair
base-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
response to oxidative stress
intrinsic apoptotic signaling pathway in response to DNA damage
response to toxic substance
response to auditory stimulus
nucleotide-excision repair, DNA incision
multicellular organism growth
global genome nucleotide-excision repair
UV-damage excision repair
nucleotide-excision repair involved in interstrand cross-link repair
damaged DNA binding
protein binding
protein domain specific binding
protein homodimerization activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09017391.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017391.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017391.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017391.hg.1 | TC09001397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000485042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000485042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000485042.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017394.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017394.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017394.hg.1 | TC09001397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017390.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017390.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017390.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017390.hg.1 | TC09001397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017393.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017393.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017393.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017393.hg.1 | TC09001397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017388.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017388.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017388.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017388.hg.1 | TC09001397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017389.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017389.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017389.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09017389.hg.1 | TC09001397.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174632.hg.1 | TC0900010950.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174631.hg.1 | TC0900010950.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174637.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174637.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174637.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174637.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174637.hg.1 | TC0900010950.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000485042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000485042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000485042.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010950.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | ENST00000496104.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174633.hg.1 | TC0900010950.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | ENST00000462523.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174630.hg.1 | TC0900010950.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | ENST00000375128.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | NM_000380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | NR_027302.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900174636.hg.1 | TC0900010950.hg.1 | Clariom D TC |

XPA
Genomic Map