Assay Details
| Gene Symbol: HTR2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000868.3 | NP_000859.1 | 3 - 4 | 769 | 93 | |
| NM_001256760.2 | NP_001243689.1 | 4 - 5 | 860 | 93 | ||
| NM_001256761.2 | NP_001243690.1 | 3 - 4 | 769 | 93 | ||
| GenBank mRNA | AK295753.1 | - | 3 - 4 | 761 | 93 | |
| AK308766.1 | - | 3 - 4 | 348 | 93 | ||
| BC028688.1 | - | 4 - 5 | 722 | 93 | ||
| BC095543.1 | - | 1 - 2 | 42 | 6519218 | 93 | |
| M81778.1 | - | 4 - 5 | 828 | 93 | ||
| U49516.1 | - | 3 - 4 | 769 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000555.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK295753.1,BC095543.1,M81778.1,NM_000868.3,NM_001256760.2,NM_001256761.2,U49516.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
behavioral fear response
cGMP biosynthetic process
cellular calcium ion homeostasis
phospholipase C-activating G-protein coupled receptor signaling pathway
phospholipase C-activating serotonin receptor signaling pathway
serotonin receptor signaling pathway
chemical synaptic transmission
locomotory behavior
feeding behavior
positive regulation of phosphatidylinositol biosynthetic process
regulation of neurological system process
regulation of appetite
response to drug
regulation of corticotropin-releasing hormone secretion
positive regulation of fat cell differentiation
release of sequestered calcium ion into cytosol
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of ERK1 and ERK2 cascade
Gq/11-coupled serotonin receptor activity
G-protein coupled serotonin receptor activity
drug binding
serotonin binding
1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding
behavioral fear response
cGMP biosynthetic process
cellular calcium ion homeostasis
phospholipase C-activating G-protein coupled receptor signaling pathway
phospholipase C-activating serotonin receptor signaling pathway
serotonin receptor signaling pathway
chemical synaptic transmission
locomotory behavior
feeding behavior
positive regulation of phosphatidylinositol biosynthetic process
regulation of neurological system process
regulation of appetite
response to drug
regulation of corticotropin-releasing hormone secretion
positive regulation of fat cell differentiation
release of sequestered calcium ion into cytosol
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
positive regulation of ERK1 and ERK2 cascade
Gq/11-coupled serotonin receptor activity
G-protein coupled serotonin receptor activity
drug binding
serotonin binding
1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007420.hg.1 | ENST00000276198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007420.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007420.hg.1 | ENST00000371951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007420.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007420.hg.1 | TC0X000555.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007421.hg.1 | ENST00000276198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007421.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007421.hg.1 | ENST00000371951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007421.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007421.hg.1 | TC0X000555.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155823.hg.1 | TC0X00008178.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008178.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | ENST00000276198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | ENST00000371950.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | ENST00000371951.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | NM_000868.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | NM_001256760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | NM_001256761.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155824.hg.1 | TC0X00008178.hg.1 | Clariom D TC |

HTR2C
Genomic Map