| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022448.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022448.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022448.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022448.hg.1 |
TC08001706.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022453.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022453.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022453.hg.1 |
TC08001706.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022452.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022452.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022452.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022452.hg.1 |
TC08001706.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800182229.hg.1 |
TC0800012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800182233.hg.1 |
TC0800012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800182235.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182235.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800182235.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800182235.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800182235.hg.1 |
TC0800012088.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000301263.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518469.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
ENST00000518884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012088.hg.1 |
NM_003695.2 |
Refseq |