| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000253807.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000289269.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000289272.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000307360.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000394633.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000398631.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000398640.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000409494.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000504120.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000506939.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000512229.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000522353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000525929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000526136.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000527624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000529310.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000529619.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000529859.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000530339.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000531613.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
ENST00000532602.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018898.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018899.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018900.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018901.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018902.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018903.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018904.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018905.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018906.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018907.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018908.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018909.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018910.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_018911.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_031411.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_031849.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_031857.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
NM_031860.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05010716.hg.1 |
TC05000737.hg.1 |
HTA TC |