Assay Details
| Gene Symbol: LDB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003893.4 | NP_003884.1 | 1 - 2 | 561 | 94 | |
| GenBank mRNA | AF064491.1 | - | 1 - 2 | 230 | 93 | |
| AF068652.1 | - | 1 - 2 | 201 | 94 | ||
| AK300588.1 | - | 1 - 2 | 428 | 94 | ||
| AK308481.1 | - | 1 - 2 | 215 | 94 | ||
| BC000482.2 | - | 1 - 2 | 540 | 2964724 | 94 | |
| BC009246.2 | - | 1 - 2 | 540 | 2964724 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001617.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF064491.1,AF068652.1,AK300588.1,BC000482.2,BC009246.2,NM_003893.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery
gastrulation with mouth forming second
hair follicle development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
anterior/posterior axis specification
epithelial structure maintenance
Wnt signaling pathway
cerebellar Purkinje cell differentiation
cellular component assembly
neuron differentiation
regulation of cell migration
regulation of DNA-templated transcription, elongation
somatic stem cell population maintenance
regulation of kinase activity
histone H3-K4 acetylation
negative regulation of erythrocyte differentiation
positive regulation of cell adhesion
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hemoglobin biosynthetic process
regulation of focal adhesion assembly
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
chromatin binding
transcription corepressor activity
protein binding
enzyme binding
LIM domain binding
protein homodimerization activity
protein self-association
transcription-dependent tethering of RNA polymerase II gene DNA at nuclear periphery
gastrulation with mouth forming second
hair follicle development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
anterior/posterior axis specification
epithelial structure maintenance
Wnt signaling pathway
cerebellar Purkinje cell differentiation
cellular component assembly
neuron differentiation
regulation of cell migration
regulation of DNA-templated transcription, elongation
somatic stem cell population maintenance
regulation of kinase activity
histone H3-K4 acetylation
negative regulation of erythrocyte differentiation
positive regulation of cell adhesion
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of hemoglobin biosynthetic process
regulation of focal adhesion assembly
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
chromatin binding
transcription corepressor activity
protein binding
enzyme binding
LIM domain binding
protein homodimerization activity
protein self-association
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10021053.hg.1 | ENST00000361198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021053.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021053.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021053.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021053.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021053.hg.1 | TC10001617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021054.hg.1 | ENST00000361198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021054.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021054.hg.1 | TC10001617.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000179880.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179880.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179880.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179880.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179880.hg.1 | TC1000011669.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000461873.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011669.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | ENST00000361198.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | ENST00000425280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | ENST00000490751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | NM_001113407.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | NM_003893.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000179879.hg.1 | TC1000011669.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 180845 | - | - |

LDB1
Genomic Map