Assay Details
| Gene Symbol: GRK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005160.3 | NP_005151.2 | 11 - 12 | 1102 | 93 | |
| XM_011529975.2 | XP_011528277.1 | 10 - 11 | 812 | 93 | ||
| GenBank mRNA | AK058103.1 | - | 10 - 11 | 892 | 93 | |
| AK292234.1 | - | 11 - 12 | 968 | 93 | ||
| AK310398.1 | - | 11 - 12 | 970 | 93 | ||
| BC036797.1 | - | 12 - 13 | 1046 | 5590378 | 93 | |
| X69117.1 | - | 11 - 12 | 955 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000165.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22003101.hg.1 | ENST00000324198.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003101.hg.1 | ENST00000545323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003101.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22003101.hg.1 | TC22000165.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | TC2200006935.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200148170.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000324198.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000418510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000455558.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | ENST00000619906.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200006935.hg.1 | NM_005160.3 | Refseq |

GRK3
Genomic Map