Assay Details
| Gene Symbol: RALBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | L42542.1 | - | 1 - 2 | 169 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the L42542.1 transcripts.
Set Membership: |
|
Panther Classification:
Molecular Function -
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009621.hg.1 | ENST00000493579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009621.hg.1 | ENST00000493579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000019317.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000383432.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000458039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000577221.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | ENST00000585015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800006639.hg.1 | NM_006788.3 | Refseq |

RALBP1
Genomic Map