Assay Details
| Gene Symbol: SIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014450.2 | NP_055265.1 | 1 - 2 | 201 | 88 | |
| GenBank mRNA | AJ010059.1 | - | 1 - 2 | 191 | 88 | |
| AK314758.1 | - | 1 - 2 | 182 | 88 | ||
| BC102029.1 | - | 1 - 2 | 199 | 40031396 | 88 | |
| BC104491.1 | - | 1 - 2 | 199 | 40031403 | 88 | |
| BC107484.1 | - | 1 - 2 | 199 | 40031398 | 88 | |
| JQ924063.1 | - | 1 - 2 | 191 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001055.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09014007.hg.1 | ENST00000259608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014007.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014007.hg.1 | TC09001055.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014010.hg.1 | ENST00000259608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014010.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014010.hg.1 | TC09001055.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014009.hg.1 | ENST00000259608.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014009.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014009.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09014009.hg.1 | TC09001055.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168373.hg.1 | TC0900009946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168374.hg.1 | TC0900009946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168372.hg.1 | TC0900009946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900168371.hg.1 | TC0900009946.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000259608.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000474403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000486859.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | ENST00000618781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009946.hg.1 | NM_014450.2 | Refseq |

SIT1
Genomic Map