Assay Details
| Gene Symbol: NEURL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004210.4 | NP_004201.3 | 1 - 2 | 499 | 101 | |
| GenBank mRNA | AF029729.1 | - | 1 - 2 | 150 | 101 | |
| BC026336.1 | - | 1 - 2 | 186 | 4812302 | 101 | |
| U87864.1 | - | 1 - 2 | 499 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000775.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translation
protein monoubiquitination
Notch signaling pathway
sperm axoneme assembly
nervous system development
brain development
skeletal muscle tissue development
lactation
negative regulation of cell proliferation
sperm motility
positive regulation of apoptotic process
positive regulation of epidermal growth factor-activated receptor activity
negative regulation of Notch signaling pathway
positive regulation of long-term neuronal synaptic plasticity
positive regulation of filopodium assembly
positive regulation of dendritic spine development
cellular response to amino acid stimulus
positive regulation of synapse maturation
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
translation factor activity, non-nucleic acid binding
ubiquitin protein ligase activity
regulation of translation
protein monoubiquitination
Notch signaling pathway
sperm axoneme assembly
nervous system development
brain development
skeletal muscle tissue development
lactation
negative regulation of cell proliferation
sperm motility
positive regulation of apoptotic process
positive regulation of epidermal growth factor-activated receptor activity
negative regulation of Notch signaling pathway
positive regulation of long-term neuronal synaptic plasticity
positive regulation of filopodium assembly
positive regulation of dendritic spine development
cellular response to amino acid stimulus
positive regulation of synapse maturation
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
translation factor activity, non-nucleic acid binding
ubiquitin protein ligase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000465048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000465048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | ENST00000465048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009258.hg.1 | ENST00000369777.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009258.hg.1 | ENST00000369780.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009258.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009258.hg.1 | TC10000775.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009253.hg.1 | ENST00000369780.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009253.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009253.hg.1 | TC10000775.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009254.hg.1 | ENST00000369780.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009254.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009254.hg.1 | TC10000775.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000160102.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160102.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160102.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160102.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160102.hg.1 | TC1000008776.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | TC1000008776.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160107.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000437579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000455386.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000465048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000465048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | ENST00000465048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008776.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160103.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160103.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160103.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160103.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160103.hg.1 | TC1000008776.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000160104.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160104.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160104.hg.1 | ENST00000369780.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160104.hg.1 | NM_004210.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160104.hg.1 | TC1000008776.hg.1 | Clariom D TC |

NEURL1
Genomic Map