Assay Details
| Gene Symbol: LBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004139.4 | NP_004130.2 | 1 - 2 | 231 | 113 | |
| GenBank mRNA | AF105067.1 | - | 1 - 2 | 142 | 113 | |
| AK313625.1 | - | 1 - 2 | 161 | 113 | ||
| AY344471.1 | - | 1 - 2 | 148 | 113 | ||
| BC022256.1 | - | 1 - 2 | 160 | 4767724 | 113 | |
| M35533.1 | - | 1 - 2 | 125 | 113 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000282.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
liver development
leukocyte chemotaxis involved in inflammatory response
macrophage activation involved in immune response
acute-phase response
cellular defense response
opsonization
lipopolysaccharide transport
lipopolysaccharide-mediated signaling pathway
detection of molecule of bacterial origin
response to lipopolysaccharide
negative regulation of tumor necrosis factor production
positive regulation of chemokine production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
toll-like receptor 4 signaling pathway
positive regulation of toll-like receptor 4 signaling pathway
positive regulation of tumor necrosis factor biosynthetic process
positive regulation of macrophage activation
negative regulation of growth of symbiont in host
innate immune response
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of phagocytosis, engulfment
positive regulation of respiratory burst involved in inflammatory response
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
positive regulation of neutrophil chemotaxis
regulation of membrane permeability
lipopolysaccharide binding
receptor binding
protein binding
lipoteichoic acid binding
liver development
leukocyte chemotaxis involved in inflammatory response
macrophage activation involved in immune response
acute-phase response
cellular defense response
opsonization
lipopolysaccharide transport
lipopolysaccharide-mediated signaling pathway
detection of molecule of bacterial origin
response to lipopolysaccharide
negative regulation of tumor necrosis factor production
positive regulation of chemokine production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
toll-like receptor 4 signaling pathway
positive regulation of toll-like receptor 4 signaling pathway
positive regulation of tumor necrosis factor biosynthetic process
positive regulation of macrophage activation
negative regulation of growth of symbiont in host
innate immune response
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of phagocytosis, engulfment
positive regulation of respiratory burst involved in inflammatory response
cellular response to lipopolysaccharide
cellular response to lipoteichoic acid
positive regulation of neutrophil chemotaxis
regulation of membrane permeability
lipopolysaccharide binding
receptor binding
protein binding
lipoteichoic acid binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20004012.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004012.hg.1 | ENST00000538599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004012.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004012.hg.1 | TC20000282.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004011.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004011.hg.1 | ENST00000538599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004011.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004011.hg.1 | TC20000282.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004013.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004013.hg.1 | ENST00000538599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004013.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004013.hg.1 | TC20000282.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004014.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004014.hg.1 | ENST00000538599.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004014.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20004014.hg.1 | TC20000282.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000007313.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150572.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150572.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150572.hg.1 | TC2000007313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150572.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150572.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150574.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150574.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150574.hg.1 | TC2000007313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150574.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150574.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150571.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150571.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150571.hg.1 | TC2000007313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150571.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150571.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150575.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150575.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150575.hg.1 | TC2000007313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150575.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150575.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150573.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150573.hg.1 | NM_004139.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000150573.hg.1 | TC2000007313.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000150573.hg.1 | ENST00000217407.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000150573.hg.1 | NM_004139.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 154033 | - | - |

LBP
Genomic Map